Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025949.3 | NP_080225.2 | 4 - 5 | 592 | 80 | |
GenBank mRNA | AK049349.1 | - | 2 - 3 | 343 | 80 | |
BC054113.1 | - | 2 - 3 | 265 | 30015425 | 80 | |
CN694399.1 | - | 4 - 5 | 578 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |