Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172554.2 | NP_766142.2 | 9 - 10 | 1073 | 126 | |
GenBank mRNA | AK038670.1 | - | 9 - 10 | 1017 | 126 | |
AK039575.1 | - | 9 - 10 | 1074 | 126 | ||
AK046891.1 | - | 9 - 10 | 1015 | 126 | ||
AK122406.1 | - | 9 - 10 | 1147 | 126 | ||
AK134967.1 | - | 9 - 10 | 1022 | 126 | ||
BC027101.1 | - | 9 - 10 | 961 | 126 | ||
BC058772.1 | - | 9 - 10 | 1014 | 126 | ||
BC080664.1 | - | 9 - 10 | 1012 | 126 | ||
BC117788.1 | - | 9 - 10 | 1015 | 40092137 | 126 | |
BC117789.2 | - | 9 - 10 | 1015 | 40092140 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |