Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016971.2 | NP_058667.1 | 1 - 2 | 215 | 65 | |
GenBank mRNA | BC104348.1 | - | 1 - 2 | 215 | 40049545 | 65 |
BC104349.1 | - | 1 - 2 | 215 | 40049546 | 65 | |
BC116235.2 | - | 1 - 2 | 211 | 40091712 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons |