Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vprbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015507.2 | NP_001015507.1 | 11 - 12 | 1798 | 109 | |
GenBank mRNA | AK122372.1 | - | 11 - 12 | 1761 | 109 | |
BC065119.1 | - | 11 - 12 | 1798 | 6853519 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |