Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cebpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024806.2 | NP_001019977.1 | 6 - 7 | 2254 | 72 | |
GenBank mRNA | AK145374.1 | - | 6 - 7 | 2236 | 72 | |
AK155990.1 | - | 6 - 7 | 2237 | 72 | ||
AK158771.1 | - | 6 - 7 | 2231 | 72 | ||
AK159081.1 | - | 6 - 7 | 2251 | 72 | ||
U19891.1 | - | 6 - 7 | 2225 | 72 | ||
U19892.1 | - | 6 - 7 | 1939 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |