Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143669.1 | - | 2 - 3 | 285 | 76 | |
AK153873.1 | - | 2 - 3 | 299 | 76 | ||
BC014720.1 | - | 2 - 3 | 279 | 4924951 | 76 | |
D13565.1 | - | 2 - 3 | 277 | 76 | ||
D13821.1 | - | 2 - 3 | 272 | 76 | ||
L20048.1 | - | 2 - 3 | 289 | 76 | ||
X75337.1 | - | 2 - 3 | 302 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |