Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023738.4 | NP_076227.1 | 9 - 10 | 1202 | 120 | |
GenBank mRNA | AF288694.1 | - | 4 - 5 | 464 | 120 | |
AK004894.2 | - | 9 - 10 | 1203 | 120 | ||
AK054410.1 | - | 9 - 10 | 1209 | 120 | ||
AK089235.1 | - | 9 - 10 | 1206 | 120 | ||
AK129009.1 | - | 9 - 10 | 1114 | 120 | ||
AK170421.1 | - | 9 - 10 | 1195 | 120 | ||
AK170644.1 | - | 8 - 9 | 1577 | 120 | ||
AK172467.1 | - | 8 - 9 | 1540 | 120 | ||
BC037693.1 | - | 7 - 8 | 976 | 120 | ||
BC138843.1 | - | 9 - 10 | 1119 | 8861865 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |