Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164634.1 | NP_001158106.1 | 6 - 7 | 1111 | 73 | |
NM_001164635.1 | NP_001158107.1 | 6 - 7 | 1019 | 73 | ||
NM_031194.5 | NP_112471.3 | 7 - 8 | 1191 | 73 | ||
GenBank mRNA | AB079895.1 | - | 5 - 6 | 848 | 73 | |
AF078869.1 | - | 6 - 7 | 966 | 73 | ||
AK031962.1 | - | 7 - 8 | 1157 | 73 | ||
AK044336.1 | - | 6 - 7 | 988 | 73 | ||
AK157891.1 | - | 6 - 7 | 1020 | 73 | ||
BC014762.1 | - | 6 - 7 | 997 | 4239544 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |