Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007854.3 | NP_031880.2 | 1 - 2 | 271 | 78 | |
GenBank mRNA | AK010950.1 | - | 1 - 2 | 208 | 78 | |
AK144451.1 | - | 1 - 2 | 164 | 78 | ||
AK154310.1 | - | 1 - 2 | 206 | 78 | ||
BC048958.1 | - | 1 - 2 | 417 | 6491511 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |