Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114595.1 | NP_001108067.1 | 12 - 13 | 2019 | 86 | |
NM_001114596.1 | NP_001108068.1 | 13 - 14 | 2055 | 86 | ||
NM_001114597.1 | NP_001108069.1 | 13 - 14 | 4887 | 86 | ||
NM_053252.3 | NP_444482.2 | 13 - 14 | 2364 | 86 | ||
GenBank mRNA | AF305087.1 | - | 13 - 14 | 4834 | 86 | |
AF305088.1 | - | 13 - 14 | 2308 | 86 | ||
AF305089.1 | - | 12 - 13 | 1963 | 86 | ||
AF305090.1 | - | 13 - 14 | 1999 | 86 | ||
AK087817.1 | - | 9 - 10 | 1606 | 86 | ||
AK089917.1 | - | 12 - 13 | 86 | |||
BC004660.1 | - | 13 - 14 | 2304 | 3497913 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |