Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023873.3 | NP_076362.2 | 10 - 11 | 1041 | 90 | |
GenBank mRNA | AF242319.1 | - | 10 - 11 | 1077 | 90 | |
AK133048.1 | - | 10 - 11 | 1047 | 90 | ||
BC050819.1 | - | 10 - 11 | 1033 | 6517145 | 90 | |
BC071260.1 | - | 10 - 11 | 1041 | 6336351 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |