Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025835.2 | NP_080111.1 | 3 - 4 | 72 | ||
GenBank mRNA | AF327060.1 | - | 3 - 4 | 72 | ||
AK004976.1 | - | 3 - 4 | 72 | |||
AK050366.1 | - | 3 - 4 | 72 | |||
AK134437.1 | - | 3 - 4 | 72 | |||
BC026531.1 | - | 3 - 4 | 72 | |||
BC037082.1 | - | 3 - 4 | 5098790 | 72 | ||
BC055946.1 | - | 3 - 4 | 4166700 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |