Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026763.2 | NP_081039.2 | 18 - 19 | 4638 | 131 | |
GenBank mRNA | AB370265.1 | - | 18 - 19 | 4730 | 131 | |
AM231153.1 | - | 12 - 13 | 2018 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |