Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052976.3 | NP_443208.1 | 9 - 10 | 1460 | 70 | |
GenBank mRNA | AK028822.1 | - | 9 - 10 | 944 | 70 | |
AK031419.1 | - | 9 - 10 | 1453 | 70 | ||
AK036038.1 | - | 9 - 10 | 1461 | 70 | ||
AK082551.1 | - | 9 - 10 | 1100 | 70 | ||
AK087469.1 | - | 9 - 10 | 924 | 70 | ||
BC004845.1 | - | 10 - 11 | 1058 | 2650009 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |