Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017525.1 | NP_001017525.1 | 8 - 9 | 1131 | 71 | |
NM_028709.2 | NP_082985.2 | 10 - 11 | 2984 | 71 | ||
GenBank mRNA | AK030864.1 | - | 3 - 4 | 2026 | 71 | |
AK039038.1 | - | 7 - 8 | 927 | 71 | ||
AK044022.1 | - | 8 - 9 | 1022 | 71 | ||
BC072592.1 | - | 10 - 11 | 1609 | 71 | ||
BC096535.1 | - | 10 - 11 | 2984 | 71 | ||
BC141145.1 | - | 10 - 11 | 2056 | 9055757 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |