Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017525.1 | NP_001017525.1 | 4 - 5 | 578 | 88 | |
NM_028709.2 | NP_082985.2 | 6 - 7 | 2431 | 88 | ||
GenBank mRNA | AK039038.1 | - | 3 - 4 | 374 | 88 | |
AK044022.1 | - | 4 - 5 | 469 | 88 | ||
BC072592.1 | - | 6 - 7 | 1056 | 88 | ||
BC096535.1 | - | 6 - 7 | 2431 | 88 | ||
BC141145.1 | - | 6 - 7 | 1503 | 9055757 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |