Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015800.3 | NP_056615.1 | 11 - 12 | 2512 | 65 | |
GenBank mRNA | AF168680.1 | - | 11 - 12 | 1964 | 65 | |
AK053197.1 | - | 9 - 10 | 1455 | 65 | ||
BC100348.1 | - | 11 - 12 | 2297 | 30521148 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |