Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163625.1 | NP_001157097.1 | 5 - 6 | 625 | 166 | |
GenBank mRNA | AK010729.1 | - | 3 - 4 | 289 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |