Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145452.1 | NP_001138924.1 | 15 - 16 | 2558 | 122 | |
GenBank mRNA | AI848431.1 | - | 1 - 2 | 138 | 122 | |
CK793318.1 | - | 2 - 3 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |