Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rela | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009045.4 | NP_033071.1 | 5 - 6 | 767 | 81 | |
GenBank mRNA | AK089489.1 | - | 5 - 6 | 512 | 81 | |
AK144565.1 | - | 5 - 6 | 512 | 81 | ||
AK154630.1 | - | 5 - 6 | 512 | 81 | ||
AK155321.1 | - | 5 - 6 | 778 | 81 | ||
AK155889.1 | - | 5 - 6 | 509 | 81 | ||
AK157076.1 | - | 5 - 6 | 499 | 81 | ||
AK158072.1 | - | 5 - 6 | 777 | 81 | ||
BC003818.1 | - | 5 - 6 | 526 | 3489295 | 81 | |
BC094053.1 | - | 5 - 6 | 767 | 3157927 | 81 | |
EF460348.1 | - | 2 - 3 | 171 | 81 | ||
M61909.1 | - | 5 - 6 | 482 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |