Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rela | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009045.4 | NP_033071.1 | 7 - 8 | 1012 | 67 | |
GenBank mRNA | AK089489.1 | - | 7 - 8 | 757 | 67 | |
AK144565.1 | - | 7 - 8 | 757 | 67 | ||
AK154630.1 | - | 7 - 8 | 757 | 67 | ||
AK155889.1 | - | 7 - 8 | 754 | 67 | ||
AK157076.1 | - | 7 - 8 | 744 | 67 | ||
AK158072.1 | - | 7 - 8 | 1022 | 67 | ||
AK211432.1 | - | 1 - 2 | 67 | 67 | ||
BC003818.1 | - | 7 - 8 | 771 | 3489295 | 67 | |
BC094053.1 | - | 7 - 8 | 1012 | 3157927 | 67 | |
DQ020177.1 | - | 5 - 6 | 581 | 67 | ||
M61909.1 | - | 7 - 8 | 727 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |