Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205820.1 | NP_991389.1 | 1 - 2 | 47 | 84 | |
GenBank mRNA | AK170044.1 | - | 1 - 2 | 49 | 84 | |
AY510706.1 | - | 1 - 2 | 47 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |