Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003950.2 | NP_001003950.1 | 2 - 3 | 407 | 68 | |
GenBank mRNA | AK152122.1 | - | 2 - 3 | 459 | 68 | |
BC080289.1 | - | 2 - 3 | 407 | 6816077 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |