Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142804.1 | NP_001136276.1 | 10 - 11 | 1510 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |