Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rlim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011276.3 | NP_035406.3 | 1 - 2 | 220 | 134 | |
GenBank mRNA | AK029295.1 | - | 1 - 2 | 214 | 134 | |
BY321640.1 | - | 1 - 2 | 221 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |