Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qrich1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114119.1 | NP_001107591.1 | 2 - 3 | 274 | 110 | |
GenBank mRNA | AK083901.1 | - | 1 - 2 | 1230 | 110 | |
AK151491.1 | - | 2 - 3 | 248 | 110 | ||
AK151530.1 | - | 2 - 3 | 248 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |