Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276442.1 | NP_001263371.1 | 7 - 8 | 643 | 103 | |
NM_025371.3 | NP_079647.1 | 7 - 8 | 647 | 103 | ||
GenBank mRNA | AK002423.1 | - | 7 - 8 | 633 | 103 | |
AK003703.1 | - | 7 - 8 | 648 | 103 | ||
BC005631.1 | - | 7 - 8 | 651 | 3964138 | 103 | |
CV560337.1 | - | 7 - 8 | 568 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |