Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053268.2 | NP_444498.2 | 18 - 19 | 1830 | 75 | |
GenBank mRNA | AB056433.1 | - | 18 - 19 | 1823 | 75 | |
AK034434.1 | - | 18 - 19 | 1830 | 75 | ||
AK035664.1 | - | 9 - 10 | 853 | 75 | ||
AK154499.1 | - | 18 - 19 | 1847 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |