Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038589.2 | NP_001033678.1 | 10 - 11 | 1059 | 114 | |
NM_021522.4 | NP_067497.2 | 11 - 12 | 1164 | 114 | ||
GenBank mRNA | AK011322.1 | - | 11 - 12 | 1100 | 114 | |
AK029977.1 | - | 11 - 12 | 1093 | 114 | ||
AK045909.1 | - | 11 - 12 | 1084 | 114 | ||
AK170933.1 | - | 1158 | 114 | |||
BC005571.1 | - | 11 - 12 | 1083 | 3157723 | 114 | |
BC050197.1 | - | 10 - 11 | 977 | 30006882 | 114 | |
CK621440.1 | - | 5 - 6 | 385 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |