Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021485.2 | NP_067460.1 | 9 - 10 | 802 | 99 | |
GenBank mRNA | AJ007938.1 | - | 9 - 10 | 802 | 99 | |
AK014412.1 | - | 9 - 10 | 881 | 99 | ||
AK090093.1 | - | 857 | 99 | |||
AK156361.1 | - | 9 - 10 | 855 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |