Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110791.1 | NP_001104261.1 | 4 - 5 | 786 | 115 | |
NM_011750.2 | NP_035880.2 | 4 - 5 | 786 | 115 | ||
NR_122118.1 | 3 - 4 | 710 | 115 | |||
GenBank mRNA | AK133895.1 | - | 4 - 5 | 575 | 115 | |
AK146151.1 | - | 4 - 5 | 547 | 115 | ||
AK147081.1 | - | 4 - 5 | 555 | 115 | ||
AK152914.1 | - | 3 - 4 | 488 | 115 | ||
AK154705.1 | - | 4 - 5 | 555 | 115 | ||
AK157845.1 | - | 4 - 5 | 555 | 115 | ||
BC009091.1 | - | 4 - 5 | 531 | 3157495 | 115 | |
BC055370.1 | - | 4 - 5 | 545 | 30055109 | 115 | |
X85802.1 | - | 4 - 5 | 543 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |