Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsbg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039114.1 | NP_001034203.1 | 10 - 11 | 1620 | 63 | |
GenBank mRNA | DQ250679.1 | - | 10 - 11 | 1620 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |