Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144858.2 | NP_659107.2 | 4 - 5 | 1005 | 67 | |
GenBank mRNA | AK157702.1 | - | 4 - 5 | 946 | 67 | |
AK164614.1 | - | 3 - 4 | 605 | 67 | ||
AK166183.1 | - | 4 - 5 | 899 | 67 | ||
AY040840.1 | - | 4 - 5 | 978 | 67 | ||
BC023942.1 | - | 1 - 2 | 77 | 67 | ||
BC052481.1 | - | 4 - 5 | 972 | 5401486 | 67 | |
BC053941.1 | - | 4 - 5 | 972 | 67 | ||
BC054069.1 | - | 4 - 5 | 972 | 67 | ||
CK387816.1 | - | 2 - 3 | 292 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |