Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142631.1 | NP_001136103.1 | 3 - 4 | 388 | 133 | |
NM_029254.1 | NP_083530.1 | 3 - 4 | 388 | 133 | ||
GenBank mRNA | AK014910.1 | - | 3 - 4 | 378 | 133 | |
AK077138.1 | - | 3 - 4 | 389 | 133 | ||
AY820306.1 | - | 2 - 3 | 289 | 133 | ||
AY820307.1 | - | 2 - 3 | 289 | 133 | ||
BC147706.1 | - | 3 - 4 | 366 | 9087465 | 133 | |
BC147719.1 | - | 3 - 4 | 366 | 9087478 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |