Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175639.1 | NP_783570.1 | 3 - 4 | 532 | 66 | |
GenBank mRNA | AK012043.1 | - | 3 - 4 | 517 | 66 | |
AK032907.1 | - | 3 - 4 | 506 | 66 | ||
AK129031.1 | - | 3 - 4 | 450 | 66 | ||
AK152321.1 | - | 3 - 4 | 522 | 66 | ||
BC079913.1 | - | 2 - 3 | 233 | 66 | ||
BC117724.2 | - | 4 - 5 | 737 | 66 | ||
BY736005.1 | - | 3 - 4 | 532 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |