Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100017953.mm.1 |
NM_001029878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017953.mm.1 |
NM_001029878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017953.mm.1 |
TC0100002228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017952.mm.1 |
NM_001029878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017952.mm.1 |
NM_001029878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017952.mm.1 |
TC0100002228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017950.mm.1 |
NM_001029878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017950.mm.1 |
NM_001029878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017950.mm.1 |
TC0100002228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017951.mm.1 |
NM_001029878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017951.mm.1 |
NM_001029878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017951.mm.1 |
TC0100002228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017954.mm.1 |
TC0100002228.mm.1 |
MTA TC |