Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021287.1 | NP_067262.1 | 8 - 9 | 1325 | 65 | |
GenBank mRNA | AK138560.1 | - | 8 - 9 | 958 | 65 | |
AK147671.1 | - | 8 - 9 | 1341 | 65 | ||
BC079860.1 | - | 8 - 9 | 1325 | 6834745 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |