Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019861.1 | NP_063914.1 | 12 - 13 | 1395 | 69 | |
GenBank mRNA | AF136280.1 | - | 12 - 13 | 1296 | 69 | |
AF197480.1 | - | 12 - 13 | 1340 | 69 | ||
AJ131851.1 | - | 12 - 13 | 1272 | 69 | ||
AK017474.1 | - | 12 - 13 | 1277 | 69 | ||
AK075862.1 | - | 12 - 13 | 1288 | 69 | ||
BC004054.1 | - | 9 - 10 | 866 | 69 | ||
BC051131.1 | - | 11 - 12 | 1514 | 69 | ||
BC058758.1 | - | 12 - 13 | 1286 | 6492834 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |