Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018775.4 | NP_061245.3 | 6 - 7 | 1215 | 67 | |
GenBank mRNA | AK138630.1 | - | 6 - 7 | 1213 | 67 | |
AK141520.1 | - | 6 - 7 | 1204 | 67 | ||
AK141816.1 | - | 6 - 7 | 1169 | 67 | ||
BC005421.1 | - | 3 - 4 | 550 | 67 | ||
BC065081.1 | - | 6 - 7 | 1149 | 6832495 | 67 | |
U88873.1 | - | 2 - 3 | 435 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |