Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003910.2 | NP_001003910.2 | 12 - 13 | 3265 | 72 | |
GenBank mRNA | AK166553.1 | - | 4 - 5 | 581 | 72 | |
AY651021.1 | - | 11 - 12 | 2905 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |