Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007496.2 | NP_031522.2 | 7 - 8 | 4627 | 73 | |
GenBank mRNA | D26046.1 | - | 7 - 8 | 4624 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |