Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034875.3 | NP_001030047.2 | 7 - 8 | 1070 | 86 | |
GenBank mRNA | AK143420.1 | - | 4 - 5 | 434 | 86 | |
BC147610.1 | - | 4 - 5 | 433 | 9056600 | 86 | |
BC147625.1 | - | 4 - 5 | 433 | 9056615 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |