Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016780.2 | NP_058060.2 | 7 - 8 | 1120 | 108 | |
GenBank mRNA | AF026509.1 | - | 7 - 8 | 1064 | 108 | |
AK156518.1 | - | 3 - 4 | 377 | 108 | ||
AK157958.1 | - | 7 - 8 | 1066 | 108 | ||
AK198913.1 | - | 181 | 108 | |||
BC125518.1 | - | 7 - 8 | 1120 | 40130033 | 108 | |
BC125520.1 | - | 7 - 8 | 1120 | 40130035 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |