Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085498.2 | NP_001078967.1 | 6 - 7 | 559 | 62 | |
GenBank mRNA | AK162346.1 | - | 2 - 3 | 136 | 62 | |
AK172123.1 | - | 5 - 6 | 708 | 62 | ||
BC147754.1 | - | 6 - 7 | 554 | 9056703 | 62 | |
BC147755.1 | - | 6 - 7 | 554 | 9056704 | 62 | |
BG078687.2 | - | 6 - 7 | 474 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |