Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008444.4 | NP_032470.3 | 3 - 4 | 1656 | 77 | |
GenBank mRNA | AK083700.1 | - | 3 - 4 | 1644 | 77 | |
AK122258.1 | - | 3 - 4 | 1617 | 77 | ||
AK147467.1 | - | 3 - 4 | 1650 | 77 | ||
AK148200.1 | - | 3 - 4 | 1637 | 77 | ||
AK161946.1 | - | 1 - 2 | 96 | 77 | ||
BC139388.1 | - | 2 - 3 | 1575 | 8862410 | 77 | |
BC139389.1 | - | 2 - 3 | 1575 | 8862411 | 77 | |
D26077.1 | - | 2 - 3 | 1569 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |