Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037906.2 | NP_001032995.1 | 17 - 18 | 2018 | 75 | |
GenBank mRNA | AK046127.1 | - | 1347 | 75 | ||
AY622226.1 | - | 17 - 18 | 2018 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |