Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133779.2 | NP_598540.2 | 6 - 7 | 820 | 80 | |
GenBank mRNA | AK019717.2 | - | 6 - 7 | 811 | 80 | |
AK044499.1 | - | 6 - 7 | 804 | 80 | ||
AK049171.1 | - | 6 - 7 | 814 | 80 | ||
AK049990.1 | - | 6 - 7 | 813 | 80 | ||
AK090020.1 | - | 6 - 7 | 812 | 80 | ||
AK155057.1 | - | 6 - 7 | 807 | 80 | ||
AK157226.1 | - | 6 - 7 | 800 | 80 | ||
BC016520.1 | - | 7 - 8 | 874 | 80 | ||
BC034175.1 | - | 6 - 7 | 796 | 5026752 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |