Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010309.3 | NP_034439.2 | 2 - 3 | 2433 | 74 | |
NM_019690.2 | NP_062664.2 | 3 - 4 | 1138 | 74 | ||
NM_022000.2 | NP_068840.2 | 3 - 4 | 1138 | 74 | ||
NM_201616.1 | NP_963910.1 | 2 - 3 | 420 | 74 | ||
NM_201618.1 | NP_963912.1 | 2 - 3 | 2433 | 74 | ||
NR_003258.1 | 3 - 4 | 126 | 74 | |||
GenBank mRNA | AF107848.1 | - | 3 - 4 | 1136 | 74 | |
AJ010163.1 | - | 3 - 4 | 1104 | 74 | ||
AK078037.1 | - | 2 - 3 | 434 | 74 | ||
AK132636.1 | - | 2 - 3 | 521 | 74 | ||
AK168996.1 | - | 2 - 3 | 391 | 74 | ||
AY519501.1 | - | 2 - 3 | 2433 | 74 | ||
AY519502.1 | - | 2 - 3 | 2433 | 74 | ||
AY519504.1 | - | 3 - 4 | 2528 | 74 | ||
AY682719.1 | - | 3 - 4 | 126 | 74 | ||
BC013824.1 | - | 2 - 3 | 233 | 74 | ||
BC013840.1 | - | 2 - 3 | 233 | 74 | ||
BC038067.1 | - | 3 - 4 | 432 | 74 | ||
BC062654.1 | - | 3 - 4 | 2575 | 74 | ||
BC080816.1 | - | 2 - 3 | 409 | 74 | ||
BC092055.1 | - | 2 - 3 | 224 | 74 | ||
BY718446.1 | - | 2 - 3 | 194 | 74 | ||
Y00703.1 | - | 2 - 3 | 232 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |