Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2109486 |
AK171381.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2109486 |
AK136488.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2109486 |
AK170947.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2109486 |
NM_001033435.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2109486 |
AB542953.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1991069 |
AK171381.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1991069 |
AK136488.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1991069 |
AK170947.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1991069 |
NM_001033435.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1991069 |
AB542953.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2109485 |
AK171381.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2109485 |
AK136488.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2109485 |
AK170947.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2109485 |
NM_001033435.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2109485 |
AB542953.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
AK032108.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
AK051965.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
AK053867.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
AK156120.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
AK169360.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
BC006874.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
BC047209.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
BC057919.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
BC066141.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
BC066826.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
BC108347.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
NM_145604.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464051 |
NM_178879.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479054 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
28185 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932376 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676178 |
AK084019.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676178 |
D49654.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676178 |
NM_008629.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676178 |
XM_001477169.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
257631 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
943867 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
204713 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
80285 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
271444 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
75577 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
51892 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2109486 |
AK171381.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2109486 |
AK136488.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2109486 |
AK170947.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2109486 |
NM_001033435.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2109486 |
AB542953.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1991069 |
AK171381.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1991069 |
AK136488.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1991069 |
AK170947.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1991069 |
NM_001033435.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1991069 |
AB542953.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2109485 |
AK171381.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2109485 |
AK136488.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2109485 |
AK170947.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2109485 |
NM_001033435.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2109485 |
AB542953.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103333 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103317 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103335 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103315 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103330 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103334 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103328 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103313 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103331 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103325 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103321 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103319 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103311 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28103314 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07480594 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02421098 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07480626 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P07480594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103325 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103311 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103360 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103314 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103401 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103398 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103321 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103393 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103400 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103315 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103429 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103295 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103357 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103435 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103333 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103330 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103317 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103430 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103303 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103326 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103403 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103409 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103385 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103342 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103370 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103334 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103301 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28103349 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103308 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103320 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09628789 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09343158 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08975147 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103349 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103344 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103334 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103342 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103343 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103304 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09330524 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09195333 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09195327 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103338 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09365664 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09195332 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09715786 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09106962 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103325 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103297 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08983897 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103303 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103326 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09514532 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103301 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103340 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103314 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09601754 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103335 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09195326 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103317 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103330 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103328 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103306 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103311 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103337 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28103321 |
- |
- |