Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145896.1 | NP_001139368.1 | 2 - 3 | 375 | 74 | |
NM_001145897.1 | NP_001139369.1 | 2 - 3 | 319 | 74 | ||
NM_198671.2 | NP_941073.2 | 2 - 3 | 507 | 74 | ||
GenBank mRNA | AK033263.1 | - | 2 - 3 | 321 | 74 | |
AK154829.1 | - | 2 - 3 | 354 | 74 | ||
BB624394.1 | - | 2 - 3 | 320 | 74 | ||
BC043094.1 | - | 2 - 3 | 507 | 6405464 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |